missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-Diphenylmethylpyrrolidine, 97+%, ACROS Organics™

Brand:  Acros Organics 348370050

Code : 29

Additional Details : CAS Number : 119237-64-8 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10560342

  • £602.80 / 5g


Estimated Shipment
08-06-2018
Add to basket


Specifications

Chemical Identifiers

(S)-2-Diphenylmethylpyrrolidine
97.0
≥97%
C17H19N
MFCD00799525
119237-64-8
100.0
97+%
237.34

Specifications

(S)-2-Diphenylmethylpyrrolidine
119237-64-8
100.0
Authentic
Oil
97+%
− 3.00
237.34
- 3.00 (25.00°C C=1,Chloroform)
5g
1.062
97.0
1.0620g/mL
Glass bottle
≥97%
C17H19N
1.5860 to 1.5880
MFCD00799525
Clear colorless to pale yellow
109°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • (S)-2-Diphenylmethylpyrrolidine

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS