missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (S)-2-(Boc-amino)-3-(1-trityl-4-imidazolyl)propionic acid, 95%

Brand:  Alfa Aesar™ H62727.03

Code : NEW

Additional Details : CAS Number : 32926-43-5 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15467786

  • £42.61 / 1g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

(S)-2-(Boc-amino)-3-(1-trityl-4-imidazolyl)propionic acid
497.58
732035
32926-43-5
MFCD00153307
Boc-His(Trt)-OH; Nα-Boc-N-im-trityl-L-histidine

Specifications

(S)-2-(Boc-amino)-3-(1-trityl-4-imidazolyl)propionic acid
Solid
≈130°C (dec.)
MFCD00153307
732035
32926-43-5
95%
497.58
1g
Boc-His(Trt)-OH; Nα-Boc-N-im-trityl-L-histidine
Videos
SDS