missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt, 98%, Thermo Scientific Chemicals
140.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
(S)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt is employed as a intermediate for Pharmaceutical and chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(S)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt is employed as a intermediate for Pharmaceutical and chemical research.
Solubility
Slightly soluble in water.
Notes
Store in freezer (-20°C). Keep container tightly closed. Store away from oxidizing agent.
(S)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt is employed as a intermediate for Pharmaceutical and chemical research.
Solubility
Slightly soluble in water.
Notes
Store in freezer (-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications
Color | White |
Quantity | 5 g |
Solubility Information | Slightly soluble in water. |
Formula Weight | 553.7 |
Percent Purity | 98% |
Physical Form | Powder |
Chemical Name or Material | (S)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'