missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-Azetidinecarboxylic acid, 99+%, ACROS Organics™

Brand:  Acros Organics 105140010

Code : 29

Additional Details : CAS Number : 2133-34-8 Weight : 0.05100kg

Packaging Glass bottle
Quantity 1g

 View more versions of this product

Product Code. 10731735

  • £122.40 / 1g


EU Stock 3
Add to basket

Specifications

Chemical Identifiers

(S)-(-)-2-Azetidinecarboxylic acid
99.0
≥99.0%
C4H7NO2
2133-34-8
100.0
99+%
101.1

Specifications

(S)-(-)-2-Azetidinecarboxylic acid
99.0
Authentic
≥99.0%
C4H7NO2
101.1
−125° (25°C c=1,water)
1g
2133-34-8
100.0
Crystalline Powder or Crystals
99+%
Glass bottle
-123° to -127° (25°C, 589nm) (c=1, water)
White to light beige
Videos
SDS