missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-Azetidinecarboxylic acid, 99+%, Thermo Scientific Chemicals

Catalog Number p-3790836
Click to view available options
Quantity:
50 mg
100 mg
250 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2133-34-8
C4H7NO2
101.1
IADUEWIQBXOCDZ-VKHMYHEASA-N
l-azetidine-2-carboxylic acid, s-azetidine-2-carboxylic acid, s---2-azetidinecarboxylic acid, 2s-azetidine-2-carboxylic acid, azetidyl-2-carboxylic acid, l-azetidine 2-carboxylic acid, s-2-azetidinecarboxylic acid, 2-azetidinecarboxylic acid, s, s---azetidine-2-carboxylic acid, 2-azetidinecarboxylic acid, l
16486
CHEBI:6198
(2S)-azetidine-2-carboxylic acid
C1CNC1C(=O)O
This item is not returnable. View return policy
2133-34-8
C4H7NO2
101.1
IADUEWIQBXOCDZ-VKHMYHEASA-N
l-azetidine-2-carboxylic acid, s-azetidine-2-carboxylic acid, s---2-azetidinecarboxylic acid, 2s-azetidine-2-carboxylic acid, azetidyl-2-carboxylic acid, l-azetidine 2-carboxylic acid, s-2-azetidinecarboxylic acid, 2-azetidinecarboxylic acid, s, s---azetidine-2-carboxylic acid, 2-azetidinecarboxylic acid, l
16486
CHEBI:6198
(2S)-azetidine-2-carboxylic acid
C1CNC1C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point 206°C to 207°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 50 mg
Merck Index 10,911
Specific Rotation -125
Solubility Information Solubility in water: 5% in water
Formula Weight 101.1
Percent Purity 99+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material (S)-(-)-2-Azetidinecarboxylic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'