missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-Acetoxysuccinic anhydride, 98%, Thermo Scientific™

Catalog Number p-7059305
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
59025-03-5
C6H6O5
158.109
MFCD00082708
SSWJHSASZZAIAU-BYPYZUCNSA-N
s---2-acetoxysuccinic anhydride, --o-acetyl-l-malic anhydride, s-2-acetoxysuccinic anhydride, 2,5-furandione, 3-acetyloxy dihydro-, 3s, s-acetoxysuccinic anhydride, --o-acetylmalic anhydride, o-acetyl-l-malic anhydride, s-3-acetoxy-dihydro-2,5-furandione, 2,5-dioxotetrahydro-3-furanyl acetate #, s-2,5-dioxotetrahydrofuran-3-yl acetate
11126528
[(3S)-2,5-dioxooxolan-3-yl] acetate
CC(=O)OC1CC(=O)OC1=O
This item is not returnable. View return policy
59025-03-5
C6H6O5
158.109
MFCD00082708
SSWJHSASZZAIAU-BYPYZUCNSA-N
s---2-acetoxysuccinic anhydride, --o-acetyl-l-malic anhydride, s-2-acetoxysuccinic anhydride, 2,5-furandione, 3-acetyloxy dihydro-, 3s, s-acetoxysuccinic anhydride, --o-acetylmalic anhydride, o-acetyl-l-malic anhydride, s-3-acetoxy-dihydro-2,5-furandione, 2,5-dioxotetrahydro-3-furanyl acetate #, s-2,5-dioxotetrahydrofuran-3-yl acetate
11126528
[(3S)-2,5-dioxooxolan-3-yl] acetate
CC(=O)OC1CC(=O)OC1=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 56°C to 58°C
Quantity 2 g
Sensitivity Moisture sensitive
Solubility Information Reacts in water.
Formula Weight 158.11
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (S)- (-)-2-Acetoxysuccinic anhydride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'