missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (S)-2,3-Di-(Boc-amino)propionic acid dicyclohexylammonium salt, 98%

Brand:  Alfa Aesar™ H62483.06

Code : NEW

Additional Details : CAS Number : 201472-68-6 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15407486

  • £233.20 / 5g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

(S)-2,3-Di-(Boc-amino)propionic acid dicyclohexylammonium salt
485.66
Boc-Dap(Boc)-OH.DCHA; (S)-Nα,Nγ-Di-Boc-2,3-diaminopropionic acid dicyclohexylammonium salt
201472-68-6
MFCD00238311

Specifications

(S)-2,3-Di-(Boc-amino)propionic acid dicyclohexylammonium salt
Solid
485.66
5g
201472-68-6
98%
MFCD00238311
Boc-Dap(Boc)-OH.DCHA; (S)-Nα,Nγ-Di-Boc-2,3-diaminopropionic acid dicyclohexylammonium salt
Videos
SDS