missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-(1-BOC-2-pyrrolidinyl)acetic acid, 97%, Thermo Scientific™

Catalog Number p-4089988
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
56502-01-3
C11H19NO4
229.27
MFCD01862939
GDWKIRLZWQQMIE-QMMMGPOBSA-N
boc-l-beta-homoproline, s-2-1-tert-butoxycarbonyl pyrrolidin-2-yl acetic acid, boc-beta-hopro-oh, 2s-1-tert-butoxycarbonyl pyrrolidin-2-yl acetic acid, boc-beta3-homopro-oh, s-2-1-boc-2-pyrrolidinyl acetic acid, s-1-boc-2-pyrrolidineacetic acid, 2-pyrrolidineacetic acid, 1-1,1-dimethylethoxy carbonyl-, 2s, 2-2s-1-tert-butoxycarbonylpyrrolidin-2-yl acetic acid, s-2-carboxymethyl-pyrrolidine-1-carboxylic acid tert-butyl ester
688605
2-[(2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidin-2-yl]acetic acid
CC(C)(C)OC(=O)N1CCCC1CC(=O)O
This item is not returnable. View return policy
56502-01-3
C11H19NO4
229.27
MFCD01862939
GDWKIRLZWQQMIE-QMMMGPOBSA-N
boc-l-beta-homoproline, s-2-1-tert-butoxycarbonyl pyrrolidin-2-yl acetic acid, boc-beta-hopro-oh, 2s-1-tert-butoxycarbonyl pyrrolidin-2-yl acetic acid, boc-beta3-homopro-oh, s-2-1-boc-2-pyrrolidinyl acetic acid, s-1-boc-2-pyrrolidineacetic acid, 2-pyrrolidineacetic acid, 1-1,1-dimethylethoxy carbonyl-, 2s, 2-2s-1-tert-butoxycarbonylpyrrolidin-2-yl acetic acid, s-2-carboxymethyl-pyrrolidine-1-carboxylic acid tert-butyl ester
688605
2-[(2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidin-2-yl]acetic acid
CC(C)(C)OC(=O)N1CCCC1CC(=O)O
CAS Min % 96.0
CAS Max % 100.0
Melting Point 97°C to 101°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation −36.6° to −40.6° (20°C, 589nm) (c=1, 4 in DMF)
Formula Weight 229.27
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material (S)-2-(1-BOC-2-pyrrolidinyl)acetic acid
Show More Show Less
compliance-icons
Product Identifier
  • (S)-2-(1-BOC-2-pyrrolidinyl)acetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'