missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (S)-1-[(R)-2-(Diphenylphosphino)ferrocenyl]ethylammonium tetrafluoroborate, 96%

Brand:  Alfa Aesar™ H60972.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15425505

  • £450.08 / 1g


Estimated Shipment date
from Supplier 11-05-2018
Add to basket

Specifications

Chemical Identifiers

(S)-1-[(R)-2-(Diphenylphosphino)ferrocenyl]ethylammonium tetrafluoroborate
C24H25BF4FeNP
501.08
MFCD17019303

Specifications

(S)-1-[(R)-2-(Diphenylphosphino)ferrocenyl]ethylammonium tetrafluoroborate
C24H25BF4FeNP
MFCD17019303
96%
501.08
1g
Videos
SDS