missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-1-Methoxy-2-propanol, 99%, Thermo Scientific Chemicals

Catalog Number p-3779018
Click to view available options
Quantity:
1 mL
5 mL
25 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
26550-55-0
C4H10O2
90.12
MFCD01632588
ARXJGSRGQADJSQ-BYPYZUCNSA-N
s-+-1-methoxy-2-propanol, 2s-1-methoxypropan-2-ol, s-1-methoxypropan-2-ol, s---1-methoxy-2-propanol, s-1-methoxy-2-propanol, 2-propanol, 1-methoxy-, 2s, 2s-1-methoxy-2-propanol, pubchem6715, s-1-methoxy-propan-2-ol
6993459
(2S)-1-methoxypropan-2-ol
CC(COC)O
This item is not returnable. View return policy
26550-55-0
C4H10O2
90.12
MFCD01632588
ARXJGSRGQADJSQ-BYPYZUCNSA-N
s-+-1-methoxy-2-propanol, 2s-1-methoxypropan-2-ol, s-1-methoxypropan-2-ol, s---1-methoxy-2-propanol, s-1-methoxy-2-propanol, 2-propanol, 1-methoxy-, 2s, 2s-1-methoxy-2-propanol, pubchem6715, s-1-methoxy-propan-2-ol
6993459
(2S)-1-methoxypropan-2-ol
CC(COC)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -97.0°C
Color Colorless
Density 0.9200g/mL
Boiling Point 120.0°C
Flash Point 30°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3OCH2CH(OH)CH3
Refractive Index 1.4020 to 1.4040
Quantity 1 mL
Specific Gravity 0.92
Specific Rotation Condition +20° (20°C c=10,CHCl3)
Specific Rotation + 20.00
Solubility Information Solubility in water: soluble
Formula Weight 90.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material (S)-(+)-1-Methoxy-2-propanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'