missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (S)-(+)-1-Indanyl isothiocyanate, 95%

Brand:  Alfa Aesar™ L19979.03

Code : NEW

Additional Details : CAS Number : 737000-84-9 Weight : 0.00100kg Transport : UN number : 2922 Chem class : 1b Pack group : III

Quantity 1g

Product Code. 15409119

  • £99.30 / 1g


Estimated Shipment date
from Supplier 26-04-2018
Add to basket

Specifications

Specifications

(S)-(+)-1-Indanyl isothiocyanate
95%
175.25
UN2922
1g
737000-84-9
C10H9NS
MFCD04039331
108° to 113°C (0.1mmHg)
Videos
SDS