missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-1-Cyclohexylethyl isothiocyanate, 97%, Thermo Scientific™

Catalog Number 15489119
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS 737000-92-9
Density 1.004
Molecular Formula C9H15NS
Refractive Index 1.5268
MDL Number MFCD05664050
Quantity 1 g
UN Number UN2922
Sensitivity Moisture sensitive
Synonym s-+-1-cyclohexylethyl isothiocyanate, 1s-1-isothiocyanatoethyl cyclohexane, ksc496q3l, 1s-1-cyclohexylethanisothiocyanate
InChI Key KMMWHCFHXACMCZ-UHFFFAOYNA-N
SMILES CC(N=C=S)C1CCCCC1
IUPAC Name (1-isothiocyanatoethyl)cyclohexane
Molecular Weight (g/mol) 169.29
PubChem CID 11528418
Formula Weight 169.29
Percent Purity 97%
Chemical Name or Material (S)-(+)-1-Cyclohexylethyl isothiocyanate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'