missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-1-BOC-3-methylpiperazine, 98%, ACROS Organics™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | 45.0°C to 48.5°C |
Color | White to Yellow |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Packaging | Glass bottle |
Quantity | 1 g |
Specific Rotation | −23° to −27° (20°C, 589nm) (c=2.5, MeOH) |
Formula Weight | 200.28 |
Show More |
Safety and Handling
Product Identifier
- (S)-1-Boc-3-methylpiperazine
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'