Alfa Aesar™ (S)-(+)-1-Benzyloxycarbonyl-3-hydroxypyrrolidine, 95%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H59643.03

1GR (S)-(+)-1-Benzyloxycarbonyl-3-hydroxypyrrolidine, 95% 1g

UNSPSC: 12352100

Code: NEW

Additional Details:
Additional Details: CAS Number: 100858-32-0 Weight: 0.00100kg


Quantity 1g

Product Code. 15435156

Quantity Price
1 £ 82.15 / 1g
Estimated Shipment date
from Supplier 07-02-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material (S)-(+)-1-Benzyloxycarbonyl-3-hydroxypyrrolidine
CAS 100858-32-0
Formula Weight 221.25
MDL Number MFCD07784363
UN Number UN2811
Synonym (S)-(+)-1-Z-3-pyrrolidinol; (S)-1-Carbobenzoxy-3-pyrrolidinol

Specification

Formula Weight 221.25
Melting Point 71° to 77°C
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material (S)-(+)-1-Benzyloxycarbonyl-3-hydroxypyrrolidine
Physical Form Solid
Assay 95%
MDL Number MFCD07784363
CAS 100858-32-0
UN Number UN2811
Color White
Quantity 1g
Synonym (S)-(+)-1-Z-3-pyrrolidinol; (S)-1-Carbobenzoxy-3-pyrrolidinol
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.