missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (S)-(+)-1-Benzyloxycarbonyl-3-hydroxypyrrolidine, 95%

Brand:  Alfa Aesar™ H59643.06

Code : NEW

Additional Details : CAS Number : 100858-32-0 Weight : 0.00500kg Transport : UN number : 2811 Chem class : 1 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 15445156

  • £286.94 / 5g


Estimated Shipment date
from Supplier 20-02-2018
Add to basket

Specifications

Chemical Identifiers

(S)-(+)-1-Benzyloxycarbonyl-3-hydroxypyrrolidine
221.25
UN2811
100858-32-0
MFCD07784363
(S)-(+)-1-Z-3-pyrrolidinol; (S)-1-Carbobenzoxy-3-pyrrolidinol

Specifications

(S)-(+)-1-Benzyloxycarbonyl-3-hydroxypyrrolidine
Solid
221.25
MFCD07784363
White
(S)-(+)-1-Z-3-pyrrolidinol; (S)-1-Carbobenzoxy-3-pyrrolidinol
100858-32-0
95%
71° to 77°C
UN2811
5g
Videos
SDS