missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-1-Amino-2-propanol, 98+%, Thermo Scientific Chemicals

Catalog Number 10500014
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
2799-17-9
C3H9NO
75.11
MFCD00064429
HXKKHQJGJAFBHI-UHFFFAOYNA-N
s-+-1-amino-2-propanol, s-1-aminopropan-2-ol, 2s-1-aminopropan-2-ol, +-isopropanolamine, s-1-amino-2-propanol, 2-propanol, 1-amino-, 2s, 1-amino-2-propanol #, 2-propanol, 1-amino-, s, ambotzfal1033
7311736
1-aminopropan-2-ol
CC(O)CN
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
2799-17-9
C3H9NO
75.11
MFCD00064429
HXKKHQJGJAFBHI-UHFFFAOYNA-N
s-+-1-amino-2-propanol, s-1-aminopropan-2-ol, 2s-1-aminopropan-2-ol, +-isopropanolamine, s-1-amino-2-propanol, 2-propanol, 1-amino-, 2s, 1-amino-2-propanol #, 2-propanol, 1-amino-, s, ambotzfal1033
7311736
1-aminopropan-2-ol
CC(O)CN

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Melting Point 19°C to 21°C
Color Colorless to Yellow
Density 0.9540g/mL
Boiling Point 160°C
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH(OH)CH2NH2
Refractive Index 1.4465 to 1.4495
Quantity 5 g
Beilstein 04, IV, 1665
Specific Gravity 0.954
Specific Rotation Condition + 18.00 (20.00°C c=1.8,H2O)
Specific Rotation + 18
Solubility Information Solubility in water: soluble. Other solubilities: soluble in organic solvents
Formula Weight 75.11
Percent Purity 98+%
Physical Form Liquid After Melting
Chemical Name or Material (S)-(+)-1-Amino-2-propanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'