missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-(4-Chlorophenyl)ethylamine, 95+%, (sum of isomers), 95% e.e., Thermo Scientific™

Catalog Number 11935261
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
4187-56-8
C8H10ClN
155.63
PINPOEWMCLFRRB-LURJTMIESA-N
s-1-4-chlorophenyl ethylamine, s-1-4-chlorophenyl ethanamine, 1s-1-4-chlorophenyl ethanamine, s-4-chloro-alpha-methylbenzylamine, s---1-4-chlorophenyl ethylamine, 1s-1-4-chlorophenyl ethan-1-amine, pubchem15221, s-4-chloro-, a-methylbenzylamine, 1s-1-4-chlorophenyl ethylamine, s-1-4-chloro-phenyl ethylamine
1550852
(1S)-1-(4-chlorophenyl)ethanamine
CC(C1=CC=C(C=C1)Cl)N
This item is not returnable. View return policy
4187-56-8
C8H10ClN
155.63
PINPOEWMCLFRRB-LURJTMIESA-N
s-1-4-chlorophenyl ethylamine, s-1-4-chlorophenyl ethanamine, 1s-1-4-chlorophenyl ethanamine, s-4-chloro-alpha-methylbenzylamine, s---1-4-chlorophenyl ethylamine, 1s-1-4-chlorophenyl ethan-1-amine, pubchem15221, s-4-chloro-, a-methylbenzylamine, 1s-1-4-chlorophenyl ethylamine, s-1-4-chloro-phenyl ethylamine
1550852
(1S)-1-(4-chlorophenyl)ethanamine
CC(C1=CC=C(C=C1)Cl)N

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 1.1150g/mL
Boiling Point 232.0°C
Quantity 25 g
Packaging Glass bottle
Refractive Index 1.5410 to 1.5430
Assay Percent Range 95% min. (GC)
Specific Gravity 1.115
Linear Formula ClC6H4CH(CH3)NH2
Solubility Information Solubility in water: moderately soluble
Formula Weight 155.63
Percent Purity 95+%
Chemical Name or Material (S)-1-(4-Chlorophenyl)ethylamine, 95% e.e
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • (S)-1-(4-Chlorophenyl)ethylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H330-Fatal if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'