missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
Used as a chiral Bronsted acid catalyst in the enantoselective Mannich reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used as a chiral Bronsted acid catalyst in the enantoselective Mannich reaction.
Solubility
Soluble in DMSO and methanol (hot).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
Used as a chiral Bronsted acid catalyst in the enantoselective Mannich reaction.
Solubility
Soluble in DMSO and methanol (hot).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
Specifications
Melting Point | >300°C |
Quantity | 250 mg |
Beilstein | 4787775 |
Solubility Information | Soluble in DMSO and methanol (hot). |
Optical Rotation | +605° (c=1 in Methanol) |
Formula Weight | 348.3 |
Percent Purity | ≥98% |
Chemical Name or Material | (S)-(+)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'