missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-1,1'-Bi(2-naphthyl) dimethanesulfonate, 97%, Thermo Scientific™

Catalog Number 15431996
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
871231-47-9
MFCD06798065
s-+-1,1-bi-2-naphthyl dimethanesul, r---1,1'-bi-2-naphthyl dimethanesulfonate, s-+-1,1'-bi-2-naphthyl dimethanesulfonate, r---1,1 inverted exclamation marka-bi-2-naphthyl dimethanesulfonate, s-+-1,1 inverted exclamation marka-bi-2-naphthyl dimethanesulfonate, 1'-methanesulfonyloxy-2,2'-binaphthalen-1-yl methanesulfonate
This item is not returnable. View return policy
871231-47-9
MFCD06798065
s-+-1,1-bi-2-naphthyl dimethanesul, r---1,1'-bi-2-naphthyl dimethanesulfonate, s-+-1,1'-bi-2-naphthyl dimethanesulfonate, r---1,1 inverted exclamation marka-bi-2-naphthyl dimethanesulfonate, s-+-1,1 inverted exclamation marka-bi-2-naphthyl dimethanesulfonate, 1'-methanesulfonyloxy-2,2'-binaphthalen-1-yl methanesulfonate

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 150°C to 154°C
Quantity 250 mg
Formula Weight 442.5
Percent Purity 97%
Physical Form Solid
Chemical Name or Material (S)-(+)-1,1'-Bi(2-naphthyl) dimethanesulfonate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'