missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1,1,2-Triphenyl-1,2-ethanediol, 98%, Thermo Scientific Chemicals

Catalog Number 15496658
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
108998-83-0
C20H18O2
290.362
MFCD00075492
GWVWUZJOQHWMFB-IBGZPJMESA-N
s---1,1,2-triphenylethane-1,2-diol, s---1,1,2-triphenyl-1,2-ethanediol, s-1,1,2-triphenylethane-1,2-diol, 2s-1,1,2-triphenylethane-1,2-diol, s-1,1,2-triphenyl-1,2-ethanediol, 1,2-ethanediol, 1,1,2-triphenyl-, 2s, 1,1,2-triphenyl-ethane-1,2-diol, s-1,1,2-triphenyl-ethane-1,2-diol, s-triphenyl-ethane-1,2-diol, s---triphenylethylene glycol
7000031
(2S)-1,1,2-triphenylethane-1,2-diol
C1=CC=C(C=C1)C(C(C2=CC=CC=C2)(C3=CC=CC=C3)O)O
This item is not returnable. View return policy
108998-83-0
C20H18O2
290.362
MFCD00075492
GWVWUZJOQHWMFB-IBGZPJMESA-N
s---1,1,2-triphenylethane-1,2-diol, s---1,1,2-triphenyl-1,2-ethanediol, s-1,1,2-triphenylethane-1,2-diol, 2s-1,1,2-triphenylethane-1,2-diol, s-1,1,2-triphenyl-1,2-ethanediol, 1,2-ethanediol, 1,1,2-triphenyl-, 2s, 1,1,2-triphenyl-ethane-1,2-diol, s-1,1,2-triphenyl-ethane-1,2-diol, s-triphenyl-ethane-1,2-diol, s---triphenylethylene glycol
7000031
(2S)-1,1,2-triphenylethane-1,2-diol
C1=CC=C(C=C1)C(C(C2=CC=CC=C2)(C3=CC=CC=C3)O)O

It is used as pharmaceutical and fine chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical and fine chemical intermediate.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 125°C to 127°C
Color Brown
Quantity 1 g
Beilstein 3208922
Formula Weight 290.36
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (S)-(-)-1,1,2-Triphenyl-1,2-ethanediol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'