missing translation for 'onlineSavingsMsg'
Learn More

RWJ 50271, Tocris Bioscience™

Inhibitor of LFA-1/ICAM mediated cell adhesion

Brand:  Tocris (Bio-Techne) 422750

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13434394

  • £795.27 / 50mg

Estimated Shipment date
from Supplier 21-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective inhibitor of LFA-1 (lymphocyte function-associated antigen-1) adhesion to soluble intercellular adhesion molecule (sICAM) (IC50 = 5 μM in HL60 cells).
Specifications

Chemical Identifiers

162112-37-0
410.415
chembl9453, 5-methyl-1-4-3-trifluoromethyl-phenyl-thiazol-2-yl-1h-pyrazole-4-carboxylic acid 3-hydroxy-propyl-amide, n-3-hydroxypropyl-5-methyl-1--4-3-trifluoromethyl phenyl-2-thiazolyl-1h-pyrazole-4-carboxamide
N-(3-hydroxypropyl)-5-methyl-1-[4-[3-(trifluoromethyl)phenyl]-1,3-thiazol-2-yl]pyrazole-4-carboxamide
C18H17F3N4O2S
HOFGTYCLOKDAES-UHFFFAOYSA-N
9953357
CC1=C(C=NN1C2=NC(=CS2)C3=CC(=CC=C3)C(F)(F)F)C(=O)NCCCO

Specifications

50mg
162112-37-0
chembl9453, 5-methyl-1-4-3-trifluoromethyl-phenyl-thiazol-2-yl-1h-pyrazole-4-carboxylic acid 3-hydroxy-propyl-amide, n-3-hydroxypropyl-5-methyl-1--4-3-trifluoromethyl phenyl-2-thiazolyl-1h-pyrazole-4-carboxamide
CC1=C(C=NN1C2=NC(=CS2)C3=CC(=CC=C3)C(F)(F)F)C(=O)NCCCO
410.415
410.41
RWJ 50271
C18H17F3N4O2S
HOFGTYCLOKDAES-UHFFFAOYSA-N
N-(3-hydroxypropyl)-5-methyl-1-[4-[3-(trifluoromethyl)phenyl]-1,3-thiazol-2-yl]pyrazole-4-carboxamide
9953357
>99%
Videos
Documents