missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ruthenium Red tetrahydrate, pure

Catalog Number 10773255
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
12790-48-6
Cl6H36N14O6Ru3-14
844.29
MFCD00167176
IMVKVBRYNZXYAP-UHFFFAOYSA-H
108195
azanide;oxygen(2-);ruthenium;ruthenium(5+);hexachloride;tetrahydrate
[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].O.O.O.O.[O-2].[O-2].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ru].[Ru+5].[Ru+5]
This item is not returnable. View return policy
12790-48-6
Cl6H36N14O6Ru3-14
844.29
MFCD00167176
IMVKVBRYNZXYAP-UHFFFAOYSA-H
108195
azanide;oxygen(2-);ruthenium;ruthenium(5+);hexachloride;tetrahydrate
[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].O.O.O.O.[O-2].[O-2].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ru].[Ru+5].[Ru+5]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Linear Formula [(NH3)5RuORu(NH3)4ORu(NH3)5]Cl6·4H2O
Merck Index 10, 8161
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ammonia
Lambda Max 530 to 536nm
Formula Weight 858.35
Percent Purity 34.6 to 36.1% (ruthenium)
Grade Pure
Molar Absorption Coefficient 8000L/(mole.cm) (lambda min., 0.02g/L H2O)
Packaging Glass bottle
Color Purple/Red
Quantity 5 g
Physical Form Crystalline Powder
Chemical Name or Material Ruthenium red tetrahydrate, Pure
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'