missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium, plasma standard solution, Specpure™ Ru 1000μg/mL, Thermo Scientific Chemicals

Catalog Number p-7050179
Click to view available options
Quantity:
50 mL
100 mL
500 mL
This item is not returnable. View return policy
MFCD00011207
This item is not returnable. View return policy
MFCD00011207

Ruthenium, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Concentration RuCl3 in 20% HCl
Quantity 50 mL
UN Number UN1789
Grade Specpure™
Solubility Miscible with water
Physical Form Liquid
Chemical Name or Material Ruthenium plasma standard solution, Ru 1000μg/mL

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'