missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium(IV) oxide, Electronic Grade, 99.95% (Metals basis), Alfa Aesar™, Premion™

For Semiconductor Industry Use For the Manufacture of Resistor Pastes

Brand:  Alfa Aesar 040336.02

Code : NEW

Additional Details : CAS Number : 12036-10-1 Weight : 0.00050kg Transport : UN number : 1479 Chem class : 5.1 Pack group : II

Quantity 0.5g

 View more versions of this product

Product Code. 11303908

  • £30.90 / 0.50g


EU Stock 1
Add to basket

Specifications

Chemical Identifiers

Ruthenium(IV) oxide
Electronic Grade
99.95% (metals basis)
133.07
Ruthenium dioxide
12036-10-1
Powder
RuO2
MFCD00011210

Specifications

Ruthenium(IV) oxide
Electronic Grade
99.95% (metals basis)
Black powder
MFCD00011210
Odorless
0.5g
12036-10-1
Powder
RuO2
133.07
Ruthenium dioxide
Black
Videos
SDS