missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium(III) nitrosylnitrate solution, Ru 1.5% w/v, Thermo Scientific Chemicals

Catalog Number p-7049062
Click to view available options
Quantity:
50 mL
250 mL
1000 mL
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
34513-98-9
MFCD00016313
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
34513-98-9
MFCD00016313

Ruthenium(III) nitrosyl nitrate used as a catalyst in various reactions namely hydrogenation, carbonylation, hydrosilylation, hydrofomylation, C-X coupling (eg Heck, Suzuki, Stille) and chiral catalysis reactions. It is also used in the production of thick-film resistors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Red to Brown
Linear Formula Ru(NO)(NO3)3
UN Number UN3093
Concentration Ru 1.5% w/v
Density 1.07 g/mL
Solubility Information Miscible with cold water.
Formula Weight 317.09
Odor Acidic
Quantity 50 mL
Physical Form Liquid
Chemical Name or Material Ruthenium(III) nitrosylnitrate solution
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'