missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium(III) iodide, anhydrous, Ru 20.5% min, Thermo Scientific Chemicals

Catalog Number p-7048868
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
13896-65-6
MFCD00016316
ruthenium iii iodide, ruthenium iii ; iodide, ruthenium 3+ triiodide, ruthenium iii iodide anhydrous, ruthenium iii iodide, anhydrous 1g, ruthenium iii iodide, anhydrous, ru min
This item is not returnable. View return policy
13896-65-6
MFCD00016316
ruthenium iii iodide, ruthenium iii ; iodide, ruthenium 3+ triiodide, ruthenium iii iodide anhydrous, ruthenium iii iodide, anhydrous 1g, ruthenium iii iodide, anhydrous, ru min

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 590°C (decomposition)
Physical Form Powder
Quantity 1 g
Formula Weight 481.78
Concentration or Composition (by Analyte or Components) Ru 20.5% min.
Sensitivity Hygroscopic
Chemical Name or Material Ruthenium(III) iodide, Anhydrous

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'