missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium, AAS standard solution, Specpure™ Ru 1000μg/mL, Thermo Scientific Chemicals

Catalog Number p-7080479
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy
MFCD00011207
This item is not returnable. View return policy
MFCD00011207

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Concentration Ru 1000μg/mL; RuCl3 in 20% HCl
Quantity 100 mL
UN Number UN1789
Grade Specpure™
Physical Form Liquid
Chemical Name or Material Ruthenium, AAS standard solution

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'