missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Ruthenium, 5% Palladium, 0.25% on activated carbon paste, Type 611

Brand:  Thermo Scientific Alfa Aesar H37300.22

Ruthenium, 5% Palladium, 0.25% on activated carbon paste, Type 611

Code : NEW

Additional Details : Weight : 0.10000kg

Product Code. 15439095

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.