missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium, 5% Palladium, 0.25% on activated carbon paste, Type 611, Thermo Scientific™

Catalog Number 15429095
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Gray
Physical Form Powder
Quantity 25 g
Odor Odorless
Chemical Name or Material Ruthenium, 5% Palladium, 0.25% on activated carbon paste, Type 611
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'