missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ruthenium, 5% on carbon paste, D101002-5

Brand:  Alfa Aesar™ H36850.06

Code : NEW

Additional Details : CAS Number : 7440-18-8 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15477697

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

Ruthenium
7440-18-8
5% on carbon paste, D101002-5
MFCD03458417

Specifications

Ruthenium
7440-18-8
MFCD03458417
Dark gray
5% on carbon paste, D101002-5
Solid
Odorless
5g
Videos
SDS