missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ruthenium, 5% on activated carbon powder, Type D101023-5, standard, reduced, nominally 50% water wet

Brand:  Alfa Aesar™ 045439.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15473985

  • £29.50 / 5g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

Ruthenium
5% on activated carbon powder, Type D101023-5, standard, reduced, nominally 50% water wet
MFCD03458417

Specifications

Ruthenium
MFCD03458417
Black
5% on activated carbon powder, Type D101023-5, standard, reduced, nominally 50% water wet
Odorless
5g
Videos
SDS