missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium, 5% on activated carbon powder, Type D101023-5, standard, reduced, nominally 50% water wet, Thermo Scientific™

Catalog Number p-7052207
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
7440-18-8
Ru
101.07
MFCD00011207 MFCD03458417
KJTLSVCANCCWHF-UHFFFAOYSA-N
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
23950
CHEBI:30682
ruthenium
[Ru]
This item is not returnable. View return policy
7440-18-8
Ru
101.07
MFCD00011207 MFCD03458417
KJTLSVCANCCWHF-UHFFFAOYSA-N
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
23950
CHEBI:30682
ruthenium
[Ru]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Black
Quantity 5 g
Odor Odorless
Chemical Name or Material Ruthenium, 5% on activated carbon powder, Type D101023-5, standard, reduced, nominally 50% water wet
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'