missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ruthenium, 5% on activated carbon powder, reduced

Aromatic ring hydrogenation

Brand:  Alfa Aesar™ 011748.14

Code : NEW

Additional Details : CAS Number : 7440-18-8 Weight : 0.02500kg Transport : UN number : 1362 Chem class : 1 Pack group : III

Quantity 25g

 View more versions of this product

Product Code. 11361818

  • £650.90 / 25g


Estimated Shipment date
from Supplier 26-03-2018
Add to basket

Specifications

Chemical Identifiers

Ruthenium
5% on activated carbon powder, Reduced
UN1362
7440-18-8
MFCD00011207

Specifications

Ruthenium
7440-18-8
UN1362
Odorless
5% on activated carbon powder, Reduced
MFCD00011207
25g
Videos
SDS