missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ruthenium, 5% on activated carbon paste, Type 619

Brand:  Alfa Aesar™ H36004.06

Code : NEW

Additional Details : CAS Number : 7440-18-8 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15487597

  • £46.30 / 5g


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

Ruthenium
7440-18-8
5% on activated carbon paste, Type 619
MFCD03458417

Specifications

Ruthenium
7440-18-8
MFCD03458417
Dark gray
5% on activated carbon paste, Type 619
Powder
Odorless
5g
Videos
SDS