missing translation for 'onlineSavingsMsg'
Learn More

RUNX3, eFluor 660, clone: R3-5G4, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 50-9817-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15520147

  • £70.27 / 25µg


Estimated Shipment date
from Supplier 26-06-2018
Add to basket

Description

Description

Description: This R3-5G4 monoclonal antibody recognizes human Runx3, a 44kDa transcription factor that belongs to the Runt domain-containing family. Runx3 is expressed in hematopoietic cells such as T cells, B cells, and myeloid cells. This transcription factor is also expressed in non-hematopoietic tissues, including the epidermis, bone, and dorsal root ganglia. Runx3 has been established to play a critical role in thymocyte development. More specifically, Runx3 promotes CD8 + T cell development by silencing Cd4 expression. Finally, studies have demonstrated the tumor suppressive activity of in a variety of cancers (e.g., gastric and breast cancers).Applications Reported: This R3-5G4 antibody has been reported for use in immunohistochemical staining.Applications Tested: This R3-5G4 antibody has been tested by immunohistochemistry on FFPE human tissue using high pH antigen retrieval.eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.The antibody can be used at less than or equal to 10 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Runx3
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q13761
AML2, CBFA3
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry
R3-5G4
eFluor 660
RUNX3
Conjugated
RUNX3
IgG1, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
864
Videos

For Research Use Only.