Learn More
RUNX3 Mouse anti-Human, Clone: R3-5G4, eBioscience™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 14-9817-80
Code : Z2
Additional Details : Weight : 0.01000kg
Description
Description: This R3-5G4 monoclonal antibody recognizes human Runx3, a 44 kDa transcription factor that belongs to the Runt domain-containing family. Runx3 is expressed in hematopoietic cells such as T cells, B cells, and myeloid cells. This transcription factor is also expressed in non-hematopoietic tissues, including the epidermis, bone, and dorsal root ganglia. Runx3 has been established to play a critical role in thymocyte development. More specifically, Runx3 promotes CD8 + T cell development by silencing Cd4 expression. Finally, studies have demonstrated the tumor suppressive activity of in a variety of cancers (e.g., gastric and breast cancers). Applications Reported: This R3-5G4 antibody has been reported for use in western blotting, and immunohistochemical staining of formalin-fixed paraffin embedded tissue sections. Applications Tested: This R3-5G4 antibody has been tested by western blotting of the Raji cell line and by immunohistochemistry of formalin-fixed paraffin embedded human skin tissue using the IHC Antigen Retrieval Solution - High pH (cat 00-4956). This can be used at less than or equal to 5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered. This gene encodes a member of the runt domain-containing family of transcription factors. A heterodimer of this protein and a beta subunit forms a complex that binds to the core DNA sequence 5′-PYGPYGGT-3′ found in a number of enhancers and promoters, and can either activate or suppress transcription. It also interacts with other transcription factors. It functions as a tumor suppressor, and the gene is frequently deleted or transcriptionally silenced in cancer. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
Runx3 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q13761 | |
Mouse | |
25 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Immunohistochemistry (Paraffin), Western Blot | |
R3-5G4 | |
Unconjugated | |
RUNX3 | |
RUNX3 | |
Affinity chromatography | |
RUO | |
864 | |
4° C | |
Liquid |
For Research Use Only.