Learn More
RUNX3 Mouse anti-Human, Biotin, Clone: R3-5G4, eBioscience™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 13-9817-82
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Description: This R3-5G4 monoclonal antibody recognizes human Runx3, a 44 kDa transcription factor that belongs to the Runt domain-containing family. Runx3 is expressed in hematopoietic cells such as T cells, B cells, and myeloid cells. This transcription factor is also expressed in non-hematopoietic tissues, including the epidermis, bone, and dorsal root ganglia. Runx3 has been established to play a critical role in thymocyte development. More specifically, Runx3 promotes CD8 + T cell development by silencing Cd4 expression. Finally, studies have demonstrated the tumor suppressive activity of in a variety of cancers (e.g., gastric and breast cancers). Applications Reported: This R3-5G4 antibody has been reported for use in western blotting, and immunohistochemical staining of formalin-fixed paraffin embedded tissue sections. Applications Tested: This R3-5G4 antibody has been tested by immunohistochemistry on human FFPE tissue sections using high pH antigen retrieval at less than or equal to 20 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.
This gene encodes a member of the runt domain-containing family of transcription factors. A heterodimer of this protein and a beta subunit forms a complex that binds to the core DNA sequence 5′-PYGPYGGT-3′ found in a number of enhancers and promoters, and can either activate or suppress transcription. It also interacts with other transcription factors. It functions as a tumor suppressor, and the gene is frequently deleted or transcriptionally silenced in cancer. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
Runx3 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2 | |
Q13761 | |
Mouse | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Immunohistochemistry (Paraffin), Western Blot | |
R3-5G4 | |
Biotin | |
RUNX3 | |
RUNX3 | |
Affinity chromatography | |
RUO | |
864 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
For Research Use Only.