missing translation for 'onlineSavingsMsg'
Learn More

Rufinamide, Tocris Bioscience™

Prolongs inactivation of sodium channels; anticonvulsant

Brand:  Tocris (Bio-Techne) 382850

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13463664

  • £431.62 / 50mg


Estimated Shipment
04-04-2019
Add to basket

Description

Description

Board spectrum anticonvulsant. Prolongs the inactivation of sodium channels and limits the frequency of action potential firing in cultured and acutely isolated neurons. Displays anticonvulsive activity in a range of animal seizure models.
Specifications

Chemical Identifiers

106308-44-5
238.198
rufinamide, inovelon, banzel, 1-2,6-difluorobenzyl-1h-1,2,3-triazole-4-carboxamide, xilep, 1-2,6-difluorophenyl methyl-1h-1,2,3-triazole-4-carboxamide, unii-wfw942pr79, 1-2,6-difluorophenyl methyl triazole-4-carboxamide, 1h-1,2,3-triazole-4-carboxamide, 1-2,6-difluorophenyl methyl, dsstox_cid_26506
1-[(2,6-difluorophenyl)methyl]triazole-4-carboxamide
C10H8F2N4O
POGQSBRIGCQNEG-UHFFFAOYSA-N
129228
C1=CC(=C(C(=C1)F)CN2C=C(N=N2)C(=O)N)F

Specifications

Rufinamide
106308-44-5
rufinamide, inovelon, banzel, 1-2,6-difluorobenzyl-1h-1,2,3-triazole-4-carboxamide, xilep, 1-2,6-difluorophenyl methyl-1h-1,2,3-triazole-4-carboxamide, unii-wfw942pr79, 1-2,6-difluorophenyl methyl triazole-4-carboxamide, 1h-1,2,3-triazole-4-carboxamide, 1-2,6-difluorophenyl methyl, dsstox_cid_26506
C1=CC(=C(C(=C1)F)CN2C=C(N=N2)C(=O)N)F
238.198
238.19
50mg
C10H8F2N4O
POGQSBRIGCQNEG-UHFFFAOYSA-N
1-[(2,6-difluorophenyl)methyl]triazole-4-carboxamide
129228
>99%
Videos