missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Rubidium sulfate, Puratronic™, 99.975% (metals basis)

Brand:  Alfa Aesar™ 012872.04

Code : NEW

Additional Details : CAS Number : 7488-54-2 Weight : 0.00200kg

Quantity 2g

 View more versions of this product

Product Code. 11317607

  • £40.10 / 2g


Estimated Shipment date
from Supplier 07-02-2018
Add to basket

Specifications

Chemical Identifiers

Rubidium sulfate
Puratronic™
MFCD00011190
7488-54-2
267

Specifications

Rubidium sulfate
Puratronic™
99.975%
267
3.613g/mL
1.513
2g
7488-54-2
Crystalline
(metals basis)
MFCD00011190
Odorless
1050°C
Videos
SDS