missing translation for 'onlineSavingsMsg'
Learn More

Rubidium molybdenum oxide, 99.5% (Metals basis), Alfa Aesar™

Brand:  Alfa Aesar 039516.09

Code : NEW

Additional Details : CAS Number : 13718-22-4 Weight : 0.01000kg

Quantity 10g

 View more versions of this product

Product Code. 11348318

  • £75.50 / 10g


Estimated Shipment date
from Supplier 26-04-2018
Add to basket

Specifications

Chemical Identifiers

Rubidium molybdenum oxide
Powder
Rb2MoO4
MFCD00049518
13718-22-4
99.5% (metals basis)
330.88
Rubidium molybdate

Specifications

Rubidium molybdenum oxide
Powder
Rb2MoO4
330.88
Rubidium molybdate
White
13718-22-4
99.5% (metals basis)
White powder
MFCD00049518
Odorless
10g
Videos
SDS