missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rubidium hydroxide hydrate, 99% (metals basis), Thermo Scientific Chemicals
173.34 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Description
Oxidative chlorination catalyst.Rubidium hydroxide hydrate is used in fireworks in place of pure rubidium to get violet color, thereby it prevents the waste of expensive element rubidium. It acts as an intermediate in the synthesis of other rubidium compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White to Gray |
Physical Form | Crystalline |
Quantity | 25 g |
Linear Formula | RbOH·xH2O |
UN Number | UN2678 |
Merck Index | 14,8285 |
Solubility Information | Soluble in water and alcohol. |
Formula Weight | 102.48 (Anhydrous) |
Percent Purity | 99% |
Sensitivity | Hygroscopic |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'