missing translation for 'onlineSavingsMsg'
Learn More

Rubidium, 99.75% (metals basis), Thermo Scientific Chemicals

Catalog Number 11319728
Click to view available options
Quantity:
5 g

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Silver
CAS 7440-17-7
Appearance Silver-colored solid
Density 1.532g/cm3
Assay Percent Range 99.75% (Metals basis)
Molecular Formula Rb
MDL Number MFCD00134055
Quantity 5 g
Synonym unii-mlt4718tjw, mlt4718tjw, rubidio, rubidium-103, rubidium-106, atom, rubidium, isotope of mass 103, rubidium, isotope of mass 106, un1423 dangerous when wet
InChI Key IGLNJRXAVVLDKE-UHFFFAOYSA-N
SMILES [Rb]
IUPAC Name rubidium
Molecular Weight (g/mol) 85.47
PubChem CID 5357696
ChEBI CHEBI:33322
Odor Odorless
Chemical Name or Material Rubidium
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'