missing translation for 'onlineSavingsMsg'
Learn More

RU 28318, potassium salt, Tocris Bioscience™

Potent, selective mineralocorticoid receptor antagonist

Brand:  Tocris (Bio-Techne) 167250

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13469893

  • £629.16 / 50mg

Estimated Shipment date
from Supplier 27-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent and selective antagonist for the mineralocorticoid receptor (MR). Inhibits aldosterone production and secretion, and selectively decreases ex-vivo MR binding in the hippocampus of adrenalectomised rats. Also decreases blood pressure in female rats following central administration in vivo.
Specifications

Chemical Identifiers

76676-34-1
440.665
7a,17a-17-hydroxy-3-oxo-7-propylpregn-4-ene-21-carboxylic acid, potassium salt
potassium;3-[(7R,8R,9S,10R,13S,14S,17R)-17-hydroxy-10,13-dimethyl-3-oxo-7-propyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl]propanoate
C25H37KO4
HXJITUGMCJCKCE-UYOQDFFISA-M
23677972
CCCC1CC2=CC(=O)CCC2(C3C1C4CCC(C4(CC3)C)(CCC(=O)[O-])O)C.[K+]

Specifications

50mg
76676-34-1
7a,17a-17-hydroxy-3-oxo-7-propylpregn-4-ene-21-carboxylic acid, potassium salt
CCCC1CC2=CC(=O)CCC2(C3C1C4CCC(C4(CC3)C)(CCC(=O)[O-])O)C.[K+]
440.665
440.66
RU 28318, potassium salt
C25H37KO4
HXJITUGMCJCKCE-UYOQDFFISA-M
potassium;3-[(7R,8R,9S,10R,13S,14S,17R)-17-hydroxy-10,13-dimethyl-3-oxo-7-propyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl]propanoate
23677972
>98%
Videos
Documents