Learn More
Tocris Bioscience™ (RS)-(Tetrazol-5-yl)glycine
Brand:  Tocris Bioscience™ 031210
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13437543
  • £204.33 / 10mg
Call for latest availability
+44 (0)1509 555 500
Description
Description
Potent NMDA receptor agonist, approximately 20 times more active than NMDA. Also available as part of the Mixed NMDA Receptor Tocriset™ .
Specifications
Specifications
138199-51-6
C3H5N5O2
10mg
>98%
Observed MW: 143.1
Soluble to 50 mM in water