Learn More
Tocris Bioscience™ (RS)-PPG
Brand:  Tocris Bioscience™ 1220/10
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13458913
  • £157.08 / 10mg
Estimated Shipment
04-03-2019
Add to basket
Description
Description
A potent and selective group III metabotropic glutamate receptor agonist, with approximately 25-fold preference for hmGlu8; anticonvulsant and neuroprotective in vivo. Also available as part of the Group III mGlu Receptor Tocriset™ .
Specifications
Specifications
(RS)-4-Phosphonophenylglycine
C8H10NO5P
10mg
120667-15-4
Observed MW: 231.14
Soluble to 100mM in 1eq. NaOH