Learn More
(RS)-MCPG, Tocris Bioscience™
Non-selective mGlu antagonist
Brand:  Tocris (Bio-Techne) 0336/10
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13447563
  • £149.94 / 10mg
Estimated Shipment date
from Supplier 25-03-2020
Description
Description
Non-selective group I/group II metabotropic glutamate receptor antagonist.
S-enantiomer and Sodium Salt also available.
Specifications
Chemical Identifiers
146669-29-6
209.201
rs-mcpg, 4-1-amino-1-carboxyethyl benzoic acid, alpha-methyl-4-carboxyphenylglycine, alpha-amino-4-carboxy-alpha-methylbenzeneacetic acid, +/--alpha-methyl-4-carboxyphenylglycine, +/--alpha-methyl-4-carboxyphenyl glycine, benzeneacetic acid, a-amino-4-carboxy-a-methyl, benzeneacetic acid, alpha-amino-4-carboxy-alpha-methyl, benzeneacetic acid, alpha-amino-4-carboxy-alpha-methyl-, +-, +/--mcpg
4-(1-amino-1-carboxyethyl)benzoic acid
C10H11NO4
DNCAZYRLRMTVSF-UHFFFAOYSA-N
1222
CC(C1=CC=C(C=C1)C(=O)O)(C(=O)O)N
Specifications
(RS)-MCPG
C10H11NO4
DNCAZYRLRMTVSF-UHFFFAOYSA-N
4-(1-amino-1-carboxyethyl)benzoic acid
1222
>99%
146669-29-6
rs-mcpg, 4-1-amino-1-carboxyethyl benzoic acid, alpha-methyl-4-carboxyphenylglycine, alpha-amino-4-carboxy-alpha-methylbenzeneacetic acid, +/--alpha-methyl-4-carboxyphenylglycine, +/--alpha-methyl-4-carboxyphenyl glycine, benzeneacetic acid, a-amino-4-carboxy-a-methyl, benzeneacetic acid, alpha-amino-4-carboxy-alpha-methyl, benzeneacetic acid, alpha-amino-4-carboxy-alpha-methyl-, +-, +/--mcpg
CC(C1=CC=C(C=C1)C(=O)O)(C(=O)O)N
209.201
209.2