missing translation for 'onlineSavingsMsg'
Learn More

(RS)-CPP, Tocris Bioscience™

Potent NMDA antagonist

Brand:  Tocris (Bio-Techne) 0173/10

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13447453

  • £191.10 / 10mg

Estimated Shipment date
from Supplier 02-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent NMDA antagonist.

R-enantiomer also available.

Specifications

Chemical Identifiers

100828-16-8
252.207
4-3-phosphonopropyl piperazine-2-carboxylic acid, 3-2-cpp, carboxypeptidase p, rs-cpp, 3-2-carboxypiperazin-4-yl propyl-1-phosphonic acid, +/--cpp, 4-3-phosphonopropyl-2-piperazinecarboxylic acid, 3h cpp, chembl22304, dl-3-2-carboxypiperazin-4-yl-propyl-1-phosphonic acid
4-(3-phosphonopropyl)piperazine-2-carboxylic acid
C8H17N2O5P
CUVGUPIVTLGRGI-UHFFFAOYSA-N
1228
C1CN(CC(N1)C(=O)O)CCCP(=O)(O)O

Specifications

10mg
100828-16-8
4-3-phosphonopropyl piperazine-2-carboxylic acid, 3-2-cpp, carboxypeptidase p, rs-cpp, 3-2-carboxypiperazin-4-yl propyl-1-phosphonic acid, +/--cpp, 4-3-phosphonopropyl-2-piperazinecarboxylic acid, 3h cpp, chembl22304, dl-3-2-carboxypiperazin-4-yl-propyl-1-phosphonic acid
C1CN(CC(N1)C(=O)O)CCCP(=O)(O)O
252.207
252.21
(RS)-CPP
C8H17N2O5P
CUVGUPIVTLGRGI-UHFFFAOYSA-N
4-(3-phosphonopropyl)piperazine-2-carboxylic acid
1228
>98%
Videos
Documents