Learn More
(RS)-APICA, Tocris Bioscience™
Selective group II mGlu antagonist
Brand:  Tocris (Bio-Techne) 107350
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13438453
  • £531.72 / 50mg
Estimated Shipment
28-06-2019
Description
Description
Rigidified analog of MPPG (Cat. No. 0853); selective group II metabotropic glutamate receptor antagonist (IC50 = 30 μM) with no significant effect on group I and III mGlu receptors at concentrations up to 1 mM. Increases extracellular glutamate concentrations and possesses unusual inverse agonist-like action.
Specifications
Chemical Identifiers
170847-18-4
257.182
rs-apica, apica drug, 1-amino-5-phosphono-indan-1-carboxylic acid, s-mppg cyclic, rs-1-amino-5-phosphonoindan-1-carboxylic acid, 1-amino-5-phosphono-indan-1-carboxylic acid rs-apica
1-amino-5-phosphono-2,3-dihydroindene-1-carboxylic acid
C10H12NO5P
ZNQZXIHSJUDIKL-UHFFFAOYSA-N
4694355
C1CC(C2=C1C=C(C=C2)P(=O)(O)O)(C(=O)O)N
Specifications
(RS)-APICA
170847-18-4
rs-apica, apica drug, 1-amino-5-phosphono-indan-1-carboxylic acid, s-mppg cyclic, rs-1-amino-5-phosphonoindan-1-carboxylic acid, 1-amino-5-phosphono-indan-1-carboxylic acid rs-apica
C1CC(C2=C1C=C(C=C2)P(=O)(O)O)(C(=O)O)N
257.182
257.18
50mg
C10H12NO5P
ZNQZXIHSJUDIKL-UHFFFAOYSA-N
1-amino-5-phosphono-2,3-dihydroindene-1-carboxylic acid
4694355
>98%
Videos