missing translation for 'onlineSavingsMsg'
Learn More

(RS)-AMPA, Tocris Bioscience™

Selective AMPA agonist

Brand:  Tocris (Bio-Techne) 016950

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13437453

  • £646.37 / 50mg

Estimated Shipment
27-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Prototypical and defining agonist for the AMPA subgroup of ionotropic glutamate receptors.

Active Enantiomer, Inactive Enantiomer and Hydrobromide Salt also available.

Specifications

Chemical Identifiers

74341-63-2
186.167
ampa, r,s-ampa, ampa pharmaceutical, alpha-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid, rs-ampa, +/--ampa, chembl13378, d,l-alpha-amino-3-hydroxy-5-methylisoxazole-4-propionic acid, r,s-alpha-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid, 4-isoxazolepropanoic acid, alpha-amino-2,3-dihydro-5-methyl-3-oxo
CHEBI:34018
CC1=C(C(=O)NO1)CC(C(=O)O)N
C7H10N2O4
UUDAMDVQRQNNHZ-UHFFFAOYSA-N
1221
2-amino-3-(5-methyl-3-oxo-1,2-oxazol-4-yl)propanoic acid

Specifications

(RS)-AMPA
74341-63-2
ampa, r,s-ampa, ampa pharmaceutical, alpha-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid, rs-ampa, +/--ampa, chembl13378, d,l-alpha-amino-3-hydroxy-5-methylisoxazole-4-propionic acid, r,s-alpha-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid, 4-isoxazolepropanoic acid, alpha-amino-2,3-dihydro-5-methyl-3-oxo
CC1=C(C(=O)NO1)CC(C(=O)O)N
186.167
CHEBI:34018
>99%
50mg
C7H10N2O4
UUDAMDVQRQNNHZ-UHFFFAOYSA-N
2-amino-3-(5-methyl-3-oxo-1,2-oxazol-4-yl)propanoic acid
1221
186.17
Videos