Learn More
RS 79948 hydrochloride, Tocris Bioscience™
Potent, selective α2 antagonist
Brand:  Tocris (Bio-Techne) 098710
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13488233
  • £140.00 / 10mg
Estimated Shipment
21-06-2018
Add to basket
Description
Description
α2-adrenoreceptor antagonist (Kd values are; 0.18, 0.19 and 0.42 nM for α2B α2C and α2A, respectively in rat).
Specifications
Chemical Identifiers
RS 79948 hydrochloride
C19H28N2O3S·HCl
400.96
(8aR,12aS,13aS)-5,8,8a,9,10,11,12,12a,13,13a-dechydro-3-methoxy-12-(ethylsulfonyl)-6H-isoquino[2,1-g][1,6]naphthyridine hydrochloride
Specifications
RS 79948 hydrochloride
>99%
400.96
(8aR,12aS,13aS)-5,8,8a,9,10,11,12,12a,13,13a-dechydro-3-methoxy-12-(ethylsulfonyl)-6H-isoquino[2,1-g][1,6]naphthyridine hydrochloride
186002-54-0
C19H28N2O3S·HCl